Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl cinnamate, 98+%

Catalog No. aaa1290636
Click to view available options
Quantity:
100 g
500 g
2500 g

CAS: 103-36-6 | C11H12O2 | 176.215 g/mol

Ethyl cinnamate is used in medical intermediate, as a pharmaceutical intermediate and in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl cinnamate is used in medical intermediate, as a pharmaceutical intermediate and in organic synthesis.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, acids, bases and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-36-6
Molecular Formula C11H12O2
Molecular Weight (g/mol) 176.215
MDL Number MFCD00009189
InChI Key KBEBGUQPQBELIU-CMDGGOBGSA-N
Synonym ethyl cinnamate, ethyl 3-phenylacrylate, ethylcinnamate, cinnamic acid ethyl ester, ethylcinnamoate, cinnamic acid, ethyl ester, ethyl trans-cinnamate, e-ethyl cinnamate, ethyl 3-phenyl-2-propenoate, ethyl benzylideneacetate
PubChem CID 637758
ChEBI CHEBI:4895
IUPAC Name ethyl (E)-3-phenylprop-2-enoate
SMILES CCOC(=O)C=CC1=CC=CC=C1

Specifications

Melting Point 7°C
Density 1.049
Boiling Point 270°C to 272°C
Flash Point 135°C (275°F)
Assay Percent Range ≥98%
Refractive Index 1.559
Quantity 500 g
Beilstein 1238804
Merck Index 14,2299
Solubility Information Insoluble in water.
Formula Weight 176.22
Percent Purity ≥98%
Chemical Name or Material Ethyl Cinnamate
Show More Show Less
EINECSNumber 203-104-6
RTECSNumber GD9010000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.