Learn More
Ethyl cyclopropanecarboxylate, 98%, Thermo Scientific Chemicals
Description
Ethyl cyclopropanecarboxylate is a reagent used in the addition of cyclopropane. It acts as annulation reagent for the conversion of aromatics to 2-methylindanones under Friedel-Crafts conditions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ethyl cyclopropanecarboxylate is a reagent used in the addition of cyclopropane. It acts as annulation reagent for the conversion of aromatics to 2-methylindanones under Friedel-Crafts conditions.
Solubility
Not miscible in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
Specifications
Density | 0.96 |
Boiling Point | 133°C to 134°C |
Flash Point | 18°C (64°F) |
Molecular Formula | C8H15BO2 |
Refractive Index | 1.42 |
Quantity | 500 g |
UN Number | UN3272 |
Beilstein | 2039752 |
Synonym | 4,4,6-trimethyl-2-vinyl-1,3,2-dioxaborinane, vinylboronic acid 2-methyl-2,4-pentanediol ester, 1,3,2-dioxaborinane,2-ethenyl-4,4,6-trimethyl, 4,4,6-trimethyl-2-vinyl-1,3,2-dioxaborinane stabilized with thf, 2-vinyl-4,4,6-trimethyl-2-bora-1,3-dioxane, vinylboronsäure-2-methyl-2,4-pentandiolester, 1,3,2-dioxaborinane, 2-ethenyl-4,4,6-trimethyl, vinylboronsaure-2-methyl-2,4-pentandiolester, vinylboronic acid 2-methyl-2,4-pentanediol ester, contains phenothiazine as stabilizer |
Solubility Information | Not miscible in water. |
Show More |
Safety and Handling
Hazard Category | H225-H302+H312+H332 |
Hazard Statement | GHS H Statement H225 Highly flammable liquid and vapour. |
Precautionary Statement | P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P312-P330-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ESTERS, N.O.S. |
EINECSNumber | 225-010-4 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.