Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl dichlorophosphite, 98%, Thermo Scientific™

Catalog No. AC188860100
Click to view available options
Quantity:
10 mL
Packaging:
Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1498-42-6
Molecular Formula C2H5Cl2OP
Molecular Weight (g/mol) 146.93
MDL Number MFCD00000525
InChI Key PVCINRPAXRJLEP-UHFFFAOYSA-N
Synonym ethyl dichlorophosphite, ethyl phosphorodichloridite, ethyl dichlorophosphinite, ethylphosphorodichloridite, ethyl phosphorodichloridoite, acmc-20akws, dichloroethoxyphosphine, dichloro ethoxy phosphane, dichloro ethoxy phosphine, c2h5opcl2
PubChem CID 73905
IUPAC Name ethyl dichlorophosphinite
SMILES CCOP(Cl)Cl

Specifications

Color Colorless
Density 1.28g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.4630 to 1.4660 (20°C, 589nm)
Linear Formula C2H5OPCl2
Quantity 10 mL
Beilstein 01, 331
Formula Weight 146.94
Percent Purity ≥97.5% (ex Cl)
Physical Form Liquid
Chemical Name or Material Ethyl dichlorophosphite, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.