Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl difluoroacetate, 97%, Acros Organics

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
454-31-9
C4H6F2O2
124.09
MFCD00013578
GZKHDVAKKLTJPO-UHFFFAOYSA-N
ethyl difluoroacetate,acetic acid, difluoro-, ethyl ester,difluoroacetic acid ethyl ester,ethyldifluoroacetate,ethyl difluoroacetic,ethyl difluoro acetate,difluoro-acetic acid ethyl ester,ethyl difluoroacetic acid,acetic acid, 2,2-difluoro-, ethyl ester,ethyldifluoracetat
9961
ethyl 2,2-difluoroacetate
CCOC(=O)C(F)F
454-31-9
C4H6F2O2
124.09
MFCD00013578
GZKHDVAKKLTJPO-UHFFFAOYSA-N
ethyl difluoroacetate,acetic acid, difluoro-, ethyl ester,difluoroacetic acid ethyl ester,ethyldifluoroacetate,ethyl difluoroacetic,ethyl difluoro acetate,difluoro-acetic acid ethyl ester,ethyl difluoroacetic acid,acetic acid, 2,2-difluoro-, ethyl ester,ethyldifluoracetat
9961
ethyl 2,2-difluoroacetate
CCOC(=O)C(F)F
Flash Point 25°C
Specific Gravity 1.242
Infrared Spectrum Authentic
Quantity 25g
Boiling Point 97.0°C
Packaging Glass bottle
Refractive Index 1.3465 to 1.3485
Density 1.2420g/mL
Color Colorless
Assay Percent Range 96% min. (GC)
Linear Formula F2CHCO2CH2CH3
UN Number 2924
Formula Weight 124.09
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Ethyl difluoroacetate
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 207-223-4