Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Difluoroacetate 98.0+%, TCI America™
SDP

Catalog No. D249825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D249825G 25 g
D24985G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D249825G Supplier TCI America Supplier No. D249825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 454-31-9
Molecular Formula C4H6F2O2
Molecular Weight (g/mol) 124.087
MDL Number MFCD00013578
InChI Key GZKHDVAKKLTJPO-UHFFFAOYSA-N
Synonym ethyl difluoroacetate, acetic acid, difluoro-, ethyl ester, difluoroacetic acid ethyl ester, ethyldifluoroacetate, ethyl difluoroacetic, ethyl difluoro acetate, difluoro-acetic acid ethyl ester, ethyl difluoroacetic acid, acetic acid, 2,2-difluoro-, ethyl ester, ethyldifluoracetat
PubChem CID 9961
IUPAC Name ethyl 2,2-difluoroacetate
SMILES CCOC(=O)C(F)F

Specifications

Color Colorless
Boiling Point 98°C
Quantity 25 g
UN Number 2924
Formula Weight 124.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Difluoroacetate
EINECSNumber (2)-3848
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.