Alfa Aesar™ Ethyl dimethylpyruvate, 97%

Manufacturer: Alfa Aesar™ H5341314

Quantity 25g

 View more versions of this product

Catalog No. AAH5341314

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 20201-24-5
    Molecular Formula C7H12O3
    Formula Weight 144.17
    MDL Number MFCD00009122
    UN Number UN3272
    Beilstein 1756668
    Synonym Ethyl 3-methyl-2-oxobutyrate; Ketovaline ethyl ester
    Chemical Name or Material Ethyl dimethylpyruvate

    Specifications

    Formula Weight 144.17
    Boiling Point 62°C (11mmHg)
    MDL Number MFCD00009122
    UN Number UN3272
    Beilstein 1756668
    Quantity 25g
    Flash Point 43°C (109°F)
    Molecular Formula C7H12O3
    CAS 20201-24-5
    Synonym Ethyl 3-methyl-2-oxobutyrate; Ketovaline ethyl ester
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H226
    Flammable liquid and vapor.
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
    Chemical Name or Material Ethyl dimethylpyruvate
    Assay 97%

    Description

    Applications
    Ethyl dimethylpyruvate is used as an allyl halide reactant in the presence of indium to release hydroxy unsaturated carbonyl compounds. It was also used in the synthesis of (2SR,4SR)-5(S)-(N-Boc)-amino-6-cyclohexyl-4-hydoxy-2-isopropyl hexanoic acid.

    Solubility
    Slightly soluble in water.

    Notes
    Stable under recommended storage conditions. Incompatible with oxidizing agents.

    GHS H Statement
    H226
    Flammable liquid and vapor.

    GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit