Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl DL-Leucate 98.0+%, TCI America™
SDP

Catalog No. E080225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E080225G 25 g
E08025G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E080225G Supplier TCI America Supplier No. E080225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10348-47-7
Molecular Formula C8H16O3
Molecular Weight (g/mol) 160.213
InChI Key QRHOWVDPHIXNEN-UHFFFAOYSA-N
Synonym DL-Leucic Acid Ethyl Ester, Ethyl DL-2-Hydroxy-4-methylvalerate, DL-2-Hydroxy-4-methylvaleric Acid Ethyl Ester
PubChem CID 112030
IUPAC Name ethyl 2-hydroxy-4-methylpentanoate
SMILES CCOC(=O)C(CC(C)C)O

Specifications

Color Colorless
Boiling Point 185°C
Quantity 25 g
Formula Weight 160.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl DL-Leucate
EINECSNumber (2)-1430
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.