Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl dodecanoate, 98%

Catalog No. p-7081254
Click to view available options
Quantity:
100 mL
500 mL
2500 mL

CAS: 106-33-2 | C14H28O2 | 228.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-33-2
Molecular Formula C14H28O2
Molecular Weight (g/mol) 228.38
MDL Number MFCD00015065
InChI Key MMXKVMNBHPAILY-UHFFFAOYSA-N
Synonym ethyl laurate, dodecanoic acid, ethyl ester, ethyl laurinate, ethyl dodecylate, lauric acid ethyl ester, lauric acid, ethyl ester, ethyl laurate natural, dodecanoic acid ethyl ester, ethyl n-dodecanote, unii-f389d4md5k
PubChem CID 7800
ChEBI CHEBI:87427
IUPAC Name ethyl dodecanoate
SMILES CCCCCCCCCCCC(=O)OCC

Specifications

Melting Point -2°C
Density 0.862
Boiling Point 268°C to 270°C
Flash Point 138°C (280°F)
Odor Fruit-like
Refractive Index 1.4315
Quantity 100 mL
Beilstein 1769671
Merck Index 14,3818
Formula Weight 228.38
Percent Purity 98%
Chemical Name or Material Ethyl dodecanoate
Show More Show Less
EINECSNumber 203-386-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.