Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl (E)-2-cyano-3-ethoxycrotonate, 98%

Catalog No. p-7029747
Click to view available options
Quantity:
5 g
25 g

CAS: 932750-29-3 | C9H13NO3 | 183.207 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 932750-29-3
Molecular Formula C9H13NO3
Molecular Weight (g/mol) 183.207
MDL Number MFCD01743485
InChI Key YKXFOGAYPIPTKF-BQYQJAHWSA-N
Synonym ethyl 2e-2-cyano-3-ethoxybut-2-enoate, ethyl e-2-cyano-3-ethoxycrotonate, ethyl 2-cyano-3-ethoxybut-2-enoate, 2-butenoic acid, 2-cyano-3-ethoxy-, ethyl ester, ethyl e-2-cyano-3-ethoxybut-2-enoate, e-ethyl 2-cyano-3-ethoxybut-2-enoate, ethyl e-2-cyano-3-ethoxy-2-butenoate, e-2-cyano-3-ethoxycrotonic acid ethyl ester, e-2-cyano-3-ethoxy-2-butenoic acid ethyl ester
PubChem CID 6387109
IUPAC Name ethyl (E)-2-cyano-3-ethoxybut-2-enoate
SMILES CCOC(=C(C#N)C(=O)OCC)C

Specifications

Melting Point 71°C to 73°C
Quantity 5 g
UN Number UN3439
Formula Weight 183.21
Percent Purity 98%
Chemical Name or Material Ethyl (E)-2-cyano-3-ethoxycrotonate
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.