Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Ether, ACS, 98%, Spectrum™ Chemical
SDP

Catalog No. 18610431 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Steel Can
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-610-431 500 mL Steel Can
18-610-427 1 L Steel Can
18-610-430 4 L Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-610-431 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. E1015500ML
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 60-29-7 , 64-17-5 , 128-37-0

Specifications

CAS Max % 100%, <3%, <0.1%
Quantity 500 mL
Packaging Steel Can
Residue after Evaporation 1e-05
Titratable Acid 0.0002 meq/g
Boiling Point 34°C
Percent Purity 0.98
Grade Reagent
Chemical Name or Material Ethyl Ether

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.