Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Ether, GR, ACS, 98.0%, MilliporeSigma™

Catalog No. 50947934 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
Packaging:
Aluminum Drum
Steel Drum
Tinplate Metal Can
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
50-947-934 4 L Aluminum Drum
50-947-937 1 L Tinplate Metal Can
50-947-656 20 L Steel Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-947-934 Supplier MilliporeSigma Supplier No. EX01853
Add to Cart
Add to Cart

Chemical Identifiers

CAS 60-29-7
Molecular Weight (g/mol) 74.12

Specifications

Melting Point −116.3°C
Quantity 4 L
Flash Point −40°C
Packaging Aluminum Drum
Residue after Evaporation 0.001% max
Assay Percent Range 98.0% min
Vapor Pressure 587 hPa (20°C)
Boiling Point 34.6°C (1.013 hPa)
Solubility Information 69 g/L
Recommended Storage Store at 2 to 25°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.