Alfa Aesar™ Ethyl formate, 97%

Manufacturer: Alfa Aesar™ A11113AE

Quantity 100mL

 View more versions of this product

Catalog No. AAA11113AE

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 109-94-4
    Molecular Formula C3H6O2
    Linear Formula HCO2CH2CH3
    Formula Weight 74.08
    MDL Number MFCD00003294
    UN Number UN1190
    Beilstein 906769
    Merck Index 14,3807
    Synonym Formic acid ethyl ester
    Chemical Name or Material Ethyl formate

    Specifications

    Formula Weight 74.08
    Physical Form Liquid
    Assay Percent Range 97%
    Health Hazard 3 GHS P Statement
    P210-P262-P243-P305+P351+P338-P403
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not get in eyes, on skin, or on clothing.
    Take precautionary measures against static discharge.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store in a well-ventilated place.
    CAS 109-94-4
    Health Hazard 1 Danger
    Flash Point −19°C (−2°F)
    Recommended Storage 2°C to 8°C
    MDL Number MFCD00003294
    Beilstein 906769
    Merck Index 14,3807
    Boiling Point 52° to 54°C
    pH 4.1
    Health Hazard 2 GHS H Statement
    H225-H302-H332-H319-H335
    Highly flammable liquid and vapor.
    Harmful if swallowed.
    Harmful if inhaled.
    Causes serious eye irritation.
    May cause respiratory irritation.
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHYL FORMATE
    RTECS Number LQ8400000
    TSCA TSCA
    Molecular Formula C3H6O2
    UN Number UN1190
    Odor Fruit-like
    Synonym Formic acid ethyl ester
    Melting Point −81°C
    Linear Formula HCO2CH2CH3
    Quantity 100mL
    Chemical Name or Material Ethyl formate
    Assay 97%

    Description

    Applications
    Ethyl formate is used as a solvent for cellulose nitrate, cellulose acetate, greases and oils. It is also used as a flavor for lemonade and essences. It is involved in the fumigation of dried fruits, as an alternative to methyl bromide.

    Solubility
    Miscible with water, benzene, alcohol, ether, acetone and most organic solvents.

    Notes
    Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with bases and oxidizing agents.

    GHS H Statement
    H225-H302-H332-H319-H335
    Highly flammable liquid and vapor.
    Harmful if swallowed.
    Harmful if inhaled.
    Causes serious eye irritation.
    May cause respiratory irritation.

    2°C to 8°C

    GHS P Statement
    P210-P262-P243-P305+P351+P338-P403
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not get in eyes, on skin, or on clothing.
    Take precautionary measures against static discharge.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store in a well-ventilated place.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit