Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ethyl formate, 98+%, Extra Dry, AcroSeal™
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -80.0°C |
| Density | 0.9170g/mL |
| Boiling Point | 54.0°C |
| Flash Point | −20°C |
| Assay Percent Range | 98% min. (GC) |
| Packaging | AcroSeal™ Glass bottle |
| Refractive Index | 1.3580 to 1.3610 |
| Linear Formula | HCO2C2H5 |
| Quantity | 100 mL |
| Merck Index | 15,3862 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Highly flammable liquid and vapour. Harmful if inhaled. Harmful if swallowed. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Call a POISON CENTER or doctor/physician if you |
| EINECSNumber | 203-721- |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction