Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl formate, 98+%, pure, ACROS Organics™

Click to view available options
Quantity:
500mL
1L
2.5L
25L
Packaging:
Glass bottle
Metal drum
109-94-4
C3H6O2
74.08
MFCD00003294
WBJINCZRORDGAQ-UHFFFAOYSA-N
areginal,formic acid, ethyl ester,ethyl methanoate,formic acid ethyl ester,aethylformiat,ethyl formic ester,ethylformate,ethylformiaat,formic ether,ethylformiat
8025
CHEBI:52342
ethyl formate
CCOC=O
109-94-4
C3H6O2
74.08
MFCD00003294
WBJINCZRORDGAQ-UHFFFAOYSA-N
areginal,formic acid, ethyl ester,ethyl methanoate,formic acid ethyl ester,aethylformiat,ethyl formic ester,ethylformate,ethylformiaat,formic ether,ethylformiat
8025
CHEBI:52342
ethyl formate
CCOC=O
Viscosity 0.4 mPa.s (20°C)
Flash Point −20°C
Water 0.1% max.
Specific Gravity 0.917
Refractive Index 1.3580 to 1.3610
Density 0.9170g/mL
Packaging Metal drum
Quantity 25L
Melting Point -80.0°C
Color Undesignated
Infrared Spectrum Authentic
Boiling Point 54.0°C
Assay Percent Range 98+%
Linear Formula HCO2C2H5
Beilstein 02, 19
Fieser 01,380; 02,197; 04,233; 12,226
Merck Index 15, 3862
Solubility Information Solubility in water: 110g/L (18°C). Other solubilities: freely soluble in ethylacetaat, isobutylacetaat,, butylacetaat, aceton, methylisobutylketon and, methylethylketon, miscible with benzene, ether and alcohol
Formula Weight 74.08
Physical Form Liquid
Percent Purity ≥98%
Grade Pure
Chemical Name or Material Ethyl formate
Show More Show Less

GHS H Statement:
Highly flammable liquid and vapour.
Harmful if inhaled.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger