Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Heptafluorobutyrate 97.0+%, TCI America™
SDP

Catalog No. H059425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H059425G 25 g
H05945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H059425G Supplier TCI America Supplier No. H059425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 356-27-4
Molecular Formula C6H5F7O2
Molecular Weight (g/mol) 242.093
MDL Number MFCD00000434
InChI Key JVHJRIQPDBCRRE-UHFFFAOYSA-N
Synonym ethyl heptafluorobutyrate, ethyl perfluorobutyrate, heptafluorobutyric acid ethyl ester, butanoic acid, heptafluoro-, ethyl ester, ethyl heptafluorobutanoate, ethyl perfluorobutanoate, ethyl heptafluoro-n-butyrate, butyric acid, heptafluoro-, ethyl ester, butanoic acid, 2,2,3,3,4,4,4-heptafluoro-, ethyl ester, pubchem12539
PubChem CID 9644
IUPAC Name ethyl 2,2,3,3,4,4,4-heptafluorobutanoate
SMILES CCOC(=O)C(C(C(F)(F)F)(F)F)(F)F

Specifications

Color Colorless
Boiling Point 96°C
Quantity 25 g
UN Number 3272
Formula Weight 242.09
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Heptafluorobutyrate
RTECSNumber ET4200000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.