Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl heptanoate, 99%, Thermo Scientific™

Catalog No. AC310490050
Click to view available options
Quantity:
5 mL
Packaging:
Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-30-9
Molecular Formula C9H18O2
Molecular Weight (g/mol) 158.24
MDL Number MFCD00009538
InChI Key TVQGDYNRXLTQAP-UHFFFAOYSA-N
Synonym ethyl enanthate, oenanthic ether, wine oil, ethyl heptylate, grape oil, ethyl heptoate, heptanoic acid, ethyl ester, cognac oil, enanthylic ether, ethyl oenanthate
PubChem CID 7797
ChEBI CHEBI:86618
IUPAC Name ethyl heptanoate
SMILES CCCCCCC(=O)OCC

Specifications

Color Colorless
Density 0.86g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.4110 to 1.4130 (20°C, 589nm)
Linear Formula CH3(CH)5COCH5
Quantity 5 mL
Merck Index 15, 3889
Formula Weight 158.27
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material Ethyl heptanoate, 99%
Show More Show Less
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.