Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl L(-)-lactate, 97%

Catalog No. p-3736465
Click to view available options
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 687-47-8 | C5H10O3 | 118.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 687-47-8
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.13
MDL Number MFCD00004518
InChI Key LZCLXQDLBQLTDK-BYPYZUCNSA-N
Synonym ethyl l-lactate, ethyl 2s-2-hydroxypropanoate, l-ethyl lactate, l---ethyl lactate, ethyl l--lactate, ethyl s-2-hydroxypropionate, ethyl l---lactate, s-ethyl lactate, propanoic acid, 2-hydroxy-, ethyl ester, 2s
PubChem CID 92831
ChEBI CHEBI:78322
IUPAC Name ethyl (2S)-2-hydroxypropanoate
SMILES CCOC(=O)C(C)O

Specifications

Acidity 0.1% max. (as lactic acid)
Melting Point -26.0°C
Density 1.0340g/mL
Boiling Point 154.0°C
Flash Point 46°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4100 to 1.4160
Linear Formula CH3CH(OH)CO2C2H5
Quantity 250 mL
Beilstein 03,264
Fieser 17,135
Merck Index 14,3817
Specific Gravity 1.034
Specific Rotation Condition − 10.00 (20.00°C neat)
Specific Rotation − 10.00
Solubility Information Solubility in water: soluble. Other solubilities: miscible with alcohols,ketones and esters
Formula Weight 118.13
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Ethyl L(-)-lactate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
May cause respiratory irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici
EINECSNumber 211-694-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.