Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl levulinate, 98%

Catalog No. p-130877
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle

CAS: 539-88-8 | C7H12O3 | 144.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 539-88-8
Molecular Formula C7H12O3
Molecular Weight (g/mol) 144.17
MDL Number MFCD00009209
InChI Key GMEONFUTDYJSNV-UHFFFAOYSA-N
Synonym ethyl levulinate, ethyl levulate, ethyl laevulinate, levulinic acid, ethyl ester, ethyl 4-ketovalerate, ethyl 3-acetylpropionate, ethyl 4-oxovalerate, pentanoic acid, 4-oxo-, ethyl ester, ethyl ketovalerate, ethyl acetylpropanoate
PubChem CID 10883
IUPAC Name ethyl 4-oxopentanoate
SMILES CCOC(=O)CCC(C)=O

Specifications

Acidity 5mg KOH/g max.
Color Yellow
Density 1.0120g/mL
Boiling Point 206°C
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4210 to 1.423
Linear Formula CH3COCH2CH2CO2C2H5
Quantity 100 g
Beilstein 03, 675
Specific Gravity 1.012
Merck Index 15, 3874
Solubility Information Solubility in water: 40g/L(20°C). Other solubilities: miscible with alcohol
Formula Weight 144.17
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl levulinate, 98%
Show More Show Less
EINECSNumber 208-728-2
RTECSNumber OI1700000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.