Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Maltol, FCC, 99%, Spectrum™ Chemical
SDP

Catalog No. 18602541 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
25 g
Packaging:
Amber Glass Bottle

Catalog No. 18-602-541

Supplier: Spectrum Chemical Manufacturing Corporation E106125GM

Only null left
Add to Cart
Add to Cart

E1061, 4940-11-8, C7H8O3

Spectrum™ Chemical Ethyl Maltol, FCC is a good flavor and sweet synergist for food, beverages, and meat products. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 11-8-4940
Molecular Formula C7H8O3
Molecular Weight (g/mol) 140.14
InChI Key YIKYNHJUKRTCJL-UHFFFAOYSA-N
IUPAC Name 2-ethyl-3-hydroxy-4H-pyran-4-one
SMILES CCC1=C(O)C(=O)C=CO1

Specifications

CAS Max % 100%
Ignition Residue 0.2%
Packaging Amber Glass Bottle
Quantity 25 g
Percent Purity 99%
Grade FCC
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.