Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl methacrylate, 99%, stabilized

Catalog No. AC215830010
Click to view available options
Quantity:
500 g
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 97-63-2 | C6H10O2 | 114.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-63-2
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.14
MDL Number MFCD00009161
InChI Key SUPCQIBBMFXVTL-UHFFFAOYSA-N
Synonym ethyl methacrylate, ethyl 2-methylacrylate, 2-propenoic acid, 2-methyl-, ethyl ester, methacrylic acid ethyl ester, ethyl 2-methyl-2-propenoate, ethyl 2-methacrylate, rhoplex ac-33, methacrylic acid, ethyl ester, rcra waste number u118, 2-methylacrylic acid, ethyl ester
PubChem CID 7343
IUPAC Name ethyl 2-methylprop-2-enoate
SMILES CCOC(=O)C(=C)C

Specifications

Melting Point -75.0°C
Color Colorless
Density 0.9100g/mL
Boiling Point 118.0°C to 119.0°C
Flash Point 15°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4130 to 1.4150
Linear Formula H2C=C(CH3)CO2C2H5
Quantity 1 kg
Beilstein 02, 423
Specific Gravity 0.91
Solubility Information Solubility in water: 4g/L in water (20°C). Other solubilities: soluble in acetone,slightly soluble in chloroform,miscible in ether and ethanol
Stabilizer 10 to 20ppm MEHQ
Viscosity 0.63 mPa.s (20°C)
Formula Weight 114.14
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Ethyl methacrylate, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
May cause an allergic skin reaction.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Wear eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 202-597-5
RTECSNumber OZ4550000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.