Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl methyl ketone, EMD-Millipore

Catalog No. M1097090005 Shop All MilliporeSigma Products
Click to view available options
Quantity:
5 mL

Chemical Identifiers

CAS 78-93-3
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00011648
InChI Key ZWEHNKRNPOVVGH-UHFFFAOYSA-N
Synonym 2-Butanone, Methyl ethyl ketone, MEK
PubChem CID 6569
ChEBI CHEBI:28398
IUPAC Name butan-2-one
SMILES CCC(C)=O

Specifications

Packaging Glass vials
Explosion Limit 1.8 to 11.5 %(V)
Ignition Point 514°C
Vapor Pressure 105 hPa (20°C)
Density Density: 0.805g/cm3 (20°C)
Assay Percent Range ≥99.5%
Linear Formula C2H5COCH3
Quantity 5 mL
Formula Weight 72.11
Chemical Name or Material Ethyl methyl ketone
Recommended Storage Store without restrictions
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.