Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl (methylthio)acetate, 98%

Catalog No. p-7027493
Click to view available options
Quantity:
25 g
100 g

CAS: 4455-13-4 | C5H10O2S | 134.193 g/mol

Ethyl (methylthio)acetate is used as an internal standard during the analytical determination of volatile sulfur compounds in port wines. Further, it is used as food flavors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl (methylthio)acetate is used as an internal standard during the analytical determination of volatile sulfur compounds in port wines. Further, it is used as food flavors.

Solubility
Miscible with alcohol. Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong alkalis.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4455-13-4
Molecular Formula C5H10O2S
Molecular Weight (g/mol) 134.193
MDL Number MFCD00009182
InChI Key MDIAKIHKBBNYHF-UHFFFAOYSA-N
Synonym ethyl methylthio acetate, ethyl 2-methylthio acetate, acetic acid, methylthio-, ethyl ester, ethyl 2-methylthio-acetate, ethyl 2-methylsulfanyl acetate, unii-6148qqn25t, ethyl alpha-methylthio acetate, methylthio acetic acid ethyl ester, ethyl .alpha. methylthio acetate, methylsulfanyl-acetic acid ethyl ester
PubChem CID 78199
IUPAC Name ethyl 2-methylsulfanylacetate
SMILES CCOC(=O)CSC

Specifications

Density 1.06
Boiling Point 176°C
Flash Point 59°C (138°F)
Odor Characteristic
Refractive Index 1.46
Quantity 25 g
UN Number UN3272
Beilstein 1744999
Solubility Information Miscible with alcohol. Immiscible with water.
Formula Weight 134.2
Percent Purity 98%
Chemical Name or Material Ethyl (methylthio)acetate
Show More Show Less
Hazard Category H226-H315-H319
Hazard Statement GHS H Statement
H226-H319-H335
Flammable liquid and vapour.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber 224-700-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.