Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl morpholine-2-carboxylate, 95%, Thermo Scientific™

Catalog No. MO08391DA
Click to view available options
:
Amber Glass Bottle; 1g

Specifications

CAS 107904-06-3
CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Molecular Formula C7H13NO3
Quantity 1g
MDL Number MFCD08751336
Synonym ethyl 2-morpholinecarboxylate, 2-morpholinecarboxylic acid ethyl ester, 2-morpholinecarboxylicacid, ethyl ester, 2-morpholinecarboxylic acid, ethyl ester, morpholine-2-carboxylic acid ethyl ester, acmc-209xhl, 2-ethoxycarbonyl morpholine
InChI Key PTWKMUDNOPBYJO-UHFFFAOYNA-N
SMILES CCOC(=O)C1CNCCO1
IUPAC Name ethyl morpholine-2-carboxylate
Molecular Weight (g/mol) 159.19
PubChem CID 15651197
Formula Weight 159.18
Percent Purity 95%
Chemical Name or Material Ethyl morpholine-2-carboxylate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.