Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl myristate, 98+%, Acros Organics

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
124-06-1
C16H32O2
256.43
MFCD00008984
MMKRHZKQPFCLLS-UHFFFAOYSA-N
ethyl myristate,tetradecanoic acid, ethyl ester,myristic acid ethyl ester,myristic acid, ethyl ester,ethylmyristate,ethyl n-tetradecanoate,ethyl myristate natural,fema no. 2445,tetradecanoic acid ethyl ester,ethyl ester tetradecanoic acid
31283
CHEBI:84849
ethyl tetradecanoate
CCCCCCCCCCCCCC(=O)OCC
124-06-1
C16H32O2
256.43
MFCD00008984
MMKRHZKQPFCLLS-UHFFFAOYSA-N
ethyl myristate,tetradecanoic acid, ethyl ester,myristic acid ethyl ester,myristic acid, ethyl ester,ethylmyristate,ethyl n-tetradecanoate,ethyl myristate natural,fema no. 2445,tetradecanoic acid ethyl ester,ethyl ester tetradecanoic acid
31283
CHEBI:84849
ethyl tetradecanoate
CCCCCCCCCCCCCC(=O)OCC
Melting Point 11.0°C to 12.0°C
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 500g
Specific Gravity 0.861
Boiling Point 295.0°C
Color Colorless
Flash Point >110°C
Density 0.8610g/mL
Assay Percent Range 98% min. (GC)
Linear Formula CH3(CH2)12COOCH2CH3
Merck Index 15, 6419
Solubility Information Solubility in water: insoluble
Formula Weight 256.43
Physical Form Liquid, Solidifying In The Cold
Percent Purity 98+%
Chemical Name or Material Ethyl myristate
Show More Show Less

GHS H Statement:
Causes skin irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 204-675-4