Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl nitroacetate, 98%

Catalog No. p-4509636
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 626-35-7 | C4H7NO4 | 133.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 626-35-7
Molecular Formula C4H7NO4
Molecular Weight (g/mol) 133.1
InChI Key FTKASJMIPSSXBP-UHFFFAOYSA-N
Synonym ethyl nitroacetate, nitroacetic acid ethyl ester, acetic acid, nitro-, ethyl ester, nitro ethyl acetate, ethylnitroacetate, acetic acid, 2-nitro-, ethyl ester, ethylnitroacetat, ethyinitroacetate, ethyl-2-nitroacetate, zlchem 269
PubChem CID 69379
IUPAC Name ethyl 2-nitroacetate
SMILES CCOC(=O)C[N+](=O)[O-]

Specifications

Color Colorless to Yellow
Density 1.1990g/mL
Boiling Point 105.0°C to 107.0°C (25.0 mmHg)
Flash Point 92°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4235 to 1.4255
Linear Formula O2NCH2CO2C2H5
Quantity 5 g
Beilstein 02,225
Specific Gravity 1.199
Formula Weight 133.1
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl nitroacetate
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 210-944-7

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.