Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ethyl nonanoate, 97%
Click to view available options
Quantity:
100 mL
500 mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | -44°C |
Density | 0.866 |
Boiling Point | 227°C to 229°C |
Flash Point | 94°C (201°F) |
Odor | Pleasant |
Refractive Index | 1.422 |
Linear Formula | CH3(CH2)7CO2CH2CH3 |
Quantity | 100 mL |
Beilstein | 1759169 |
Merck Index | 14,3838 |
Show More |
Safety and Handling
Hazard Category | Warning |
Hazard Statement | GHS H Statement H315-H319 Causes skin irritation. Causes serious eye irritation. |
Precautionary Statement | GHS P Statement P280g-P305+P351+P338 Wear protective gloves. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. |
EINECSNumber | 204-615-7 |
RTECSNumber | RA6845000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction