Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl o-Tolylacetate 97.0+%, TCI America™
SDP

Catalog No. T122525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T122525G 25 g
T12255G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T122525G Supplier TCI America Supplier No. T122525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 40291-39-2
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.231
MDL Number MFCD00009179
InChI Key HTDQOYWJDUMNHX-UHFFFAOYSA-N
Synonym ethyl o-tolylacetate, ethyl 2-o-tolyl acetate, ethyl 2-2-methylphenyl acetate, 2-methylphenylacetic acid ethyl ester, o-tolylacetic acid ethyl ester, ethyl o-methylphenylacetate, ethyl 2-methylphenylacetate, benzeneacetic acid, 2-methyl-, ethyl ester, toluene=ethyl acetate, ethyl 2-o-tolylacetate
PubChem CID 96528
IUPAC Name ethyl 2-(2-methylphenyl)acetate
SMILES CCOC(=O)CC1=CC=CC=C1C

Specifications

Color Yellow
Boiling Point 77°C
Quantity 25 g
Formula Weight 178.23
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl o-Tolylacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.