Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl octanoate, 99%

Catalog No. AAB2345022
Click to view available options
Quantity:
100 g
500 g

CAS: 106-32-1 | C10H20O2 | 172.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-32-1
Molecular Formula C10H20O2
Molecular Weight (g/mol) 172.27
MDL Number MFCD00009552
InChI Key YYZUSRORWSJGET-UHFFFAOYSA-N
Synonym ethyl caprylate, octanoic acid, ethyl ester, ethyl n-octanoate, caprylic acid ethyl ester, ethyl octoate, ethyl octylate, octanoic acid ethyl ester, natural, unii-81c5mop582, fema no. 2449
PubChem CID 7799
ChEBI CHEBI:87426
IUPAC Name ethyl octanoate
SMILES CCCCCCCC(=O)OCC

Specifications

Melting Point -47°C
Density 0.872
Boiling Point 206°C to 208°C
Flash Point 79°C (174°F)
Odor Ester-like
Refractive Index 1.417
Linear Formula CH3(CH2)6CO2CH2CH3
Quantity 100 g
Merck Index 14,3778
Formula Weight 172.27
Percent Purity 99%
Chemical Name or Material Ethyl octanoate
Show More Show Less
Hazard Category H227-H315-H319
Precautionary Statement P210-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
EINECSNumber 203-385-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.