Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ethyl orange sodium salt, 92%, Thermo Scientific™
Click to view available options
:
10g
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Indicator
Specifications
Specifications
| CAS | 62758-12-7 |
| Assay Percent Range | 92% |
| Molecular Formula | C16H18N3NaO3S |
| MDL Number | MFCD00007503 |
| InChI Key | YZORUOZKRBVLEG-UHFFFAOYSA-M |
| SMILES | CCN(CC)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)S(=O)(=O)[O-].[Na+] |
| Infrared Spectrum | Authentic |
| IUPAC Name | sodium;4-[[4-(diethylamino)phenyl]diazenyl]benzenesulfonate |
| Molecular Weight (g/mol) | 355.388 |
| PubChem CID | 23676309 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction