Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl p-Anisate 99.0+%, TCI America™
SDP

Catalog No. A0933100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0933100ML 100 mL
A093325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0933100ML Supplier TCI America Supplier No. A0933100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 94-30-4
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00015151
InChI Key FHUODBDRWMIBQP-UHFFFAOYSA-N
Synonym ethyl p-anisate, ethyl anisate, ethyl p-methoxybenzoate, p-anisic acid, ethyl ester, benzoic acid, 4-methoxy-, ethyl ester, 4-methoxybenzoic acid ethyl ester, benzoic acid, p-methoxy-, ethyl ester, ethyl p-anisoate, unii-kj95h2s7nm, p-anisic acid ethyl ester
PubChem CID 60979
IUPAC Name ethyl 4-methoxybenzoate
SMILES CCOC(=O)C1=CC=C(C=C1)OC

Specifications

Color Colorless
Boiling Point 143°C
Quantity 100 mL
Formula Weight 180.20
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl p-Anisate
EINECSNumber (3)-3054
RTECSNumber BZ4697000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.