Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl phenoxyacetate, 98%, Thermo Scientific™

Catalog No. AC334940250
Click to view available options
Quantity:
5 g
25 g
100 g

Chemical Identifiers

CAS 2555-49-9
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.2
MDL Number MFCD00026895
InChI Key MGZFVSUXQXCEHM-UHFFFAOYSA-N
Synonym ethyl phenoxyacetate, acetic acid, phenoxy-, ethyl ester, phenoxyacetic acid ethyl ester, acetic acid, 2-phenoxy-, ethyl ester, phenoxy-acetic acid ethyl ester, phenoxyacetic acid, ethyl ester, ethyl2-phenoxyacetate, pubchem12485, acmc-1ckja, ethylphenoxyacetate
PubChem CID 17365
IUPAC Name ethyl 2-phenoxyacetate
SMILES CCOC(=O)COC1=CC=CC=C1

Specifications

Color Yellow
Density 1.1000g/mL
Boiling Point 136°C (19.0 mmHg)
Flash Point >113°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Refractive Index 1.5035
Quantity 25 g
Specific Gravity 1.1
Solubility Information Solubility in water: immiscible in water
Formula Weight 180.2
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl phenoxyacetate, 98%
Show More Show Less
EINECSNumber 219-867-3
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.