Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl potassium oxalate, 97%

Catalog No. p-7032906
Click to view available options
Quantity:
5 g
25 g

CAS: 1906-57-6 | C4H5KO4 | 156.178 g/mol

Ethyl potassium oxalate is used to prepare oxalic acid ethyl ester 2-oxo-3-phenyl-propyl ester by reacting with 1-bromo-3-phenyl-acetone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl potassium oxalate is used to prepare oxalic acid ethyl ester 2-oxo-3-phenyl-propyl ester by reacting with 1-bromo-3-phenyl-acetone.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1906-57-6
Molecular Formula C4H5KO4
Molecular Weight (g/mol) 156.178
MDL Number MFCD00045861
InChI Key RLPQQBNSTHRHEK-UHFFFAOYSA-M
Synonym ethyl potassium oxalate, potassium ethyl oxalate, ethanedioic acid, monoethyl ester, potassium salt, potassium 2-ethoxy-2-oxoacetate, oxalic acid 1-ethyl 2-potassium salt, ethanedioic acid, 1-ethyl ester, potassium salt 1:1, potassium ethoxy oxo acetate, ambkkkkk567, oxalic acid, monoethyl ester, potassium salt, potassium monoethyl oxalate
PubChem CID 23678856
IUPAC Name potassium;2-ethoxy-2-oxoacetate
SMILES CCOC(=O)C(=O)[O-].[K+]

Specifications

Melting Point 220°C to 223°C
Quantity 5 g
UN Number UN2811
Formula Weight 156.18
Percent Purity 97%
Chemical Name or Material Ethyl potassium oxalate
Hazard Category H302+H312
Hazard Statement GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 217-606-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.