Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl pyruvate, 98%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Glass bottle
617-35-6
C5H8O3
116.12
MFCD00009123
XXRCUYVCPSWGCC-UHFFFAOYSA-N
ethyl pyruvate,ethyl 2-oxopropionate,pyruvic acid, ethyl ester,propanoic acid, 2-oxo-, ethyl ester,pyruvic acid ethyl ester,ethyl pyroracemate,ethyl acetylformate,2-oxo-propionic acid ethyl ester,ethyl pyruvate natural,ethyl alpha-ketopropionate
12041
ethyl 2-oxopropanoate
CCOC(=O)C(=O)C
617-35-6
C5H8O3
116.12
MFCD00009123
XXRCUYVCPSWGCC-UHFFFAOYSA-N
ethyl pyruvate,ethyl 2-oxopropionate,pyruvic acid, ethyl ester,propanoic acid, 2-oxo-, ethyl ester,pyruvic acid ethyl ester,ethyl pyroracemate,ethyl acetylformate,2-oxo-propionic acid ethyl ester,ethyl pyruvate natural,ethyl alpha-ketopropionate
12041
ethyl 2-oxopropanoate
CCOC(=O)C(=O)C
Viscosity 1.18 mPa.s
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4040 to 1.4060
Flash Point 45°C
Specific Gravity 1.0453
Density 1.0453g/mL
Melting Point -58.0°C
Boiling Point 148.0°C to 150.0°C
Color Yellow
Quantity 100g
Assay Percent Range 98%
Linear Formula CH3COCO2C2H5
Beilstein 03, 616
Merck Index 15, 8135
Solubility Information Solubility in water: 10g/L (20°C). Other solubilities: soluble in ethanol,ether
Formula Weight 116.12
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Ethyl pyruvate, 98%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 210-511-2

TSCA : TSCA