Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl pyruvate, 98%

Manufacturer:  Alfa Aesar A1504518

Quantity 50g

 View more versions of this product

Catalog No. AAA1504518


  Check Availability
Add to cart

Specifications

Description

Solubility
Miscible with water, ethanol and ether.

Notes
Store in a cool place. Incompatible with oxidizing agents.
Description & Specifications

Chemical Identifiers

617-35-6
116.12
UN3272
14,8021
C5H8O3
MFCD00009123
1071466
Ethyl Pyruvate

Specifications

50g
1.050
C5H8O3
MFCD00009123
1071466
Ethyl Pyruvate
617-35-6
98%
116.12
UN3272
14,8021
98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H226

GHS P Statement
P210-P280-P240-P241-P233-P242-P243-P303+P361+P353-P403+P235-P501a

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.

TSCA : TSCA

storageNote1 : Store cold

Recommended Storage : 2° to 8°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit