Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Ethyl Red

Catalog No. AAA1836718
Click to view available options
Quantity:
10 g
50 g
250 g

CAS: 76058-33-8 | C17H18N3O2 | 296.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76058-33-8
Molecular Formula C17H18N3O2
Molecular Weight (g/mol) 296.35
MDL Number MFCD00002427
InChI Key HBRCDTRQDHMTDA-UHFFFAOYSA-M
Synonym 2-(4-Diethylaminophenylazo)benzoic acid
PubChem CID 54832
SMILES CCN(CC)C1=CC=C(C=C1)N=NC1=CC=CC=C1C([O-])=O

Specifications

Melting Point ∼135°C (decomposition)
Beilstein 3366056
IUPAC Name 2-[[4-(diethylamino)phenyl]diazenyl]benzoic acid
Formula Weight 297.36
Quantity 50 g
Chemical Name or Material Ethyl Red
Hazard Category H312+H332
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P271-P280-P302+P352-P304+P340-P312-P363-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.