Learn More
Ethyl thiophene-2-glyoxylate, 97%
Description
Ethyl thiophene-2-glyoxylate is used for drug discovery using chemic informatics-based design and it is used as a catalyst, synthesis of raw material.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ethyl thiophene-2-glyoxylate is used for drug discovery using chemic informatics-based design and it is used as a catalyst, synthesis of raw material.
Solubility
Difficult to mix.
Notes
Keep container tightly closed. Store away from oxidizing agents.

Specifications
Specifications
Density | 1.25 |
Boiling Point | 102°C to 110°C (0.8 mmHg) |
Flash Point | >110°C (230°F) |
Refractive Index | 1.5505 |
Quantity | 10 g |
Beilstein | 5651 |
Solubility Information | Difficult to mix. |
Formula Weight | 184.21 |
Percent Purity | 97% |
Chemical Name or Material | Ethyl thiophene-2-glyoxylate |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
EINECSNumber | 223-793-7 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.