Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl thiophene-3-carboxylate, 98%

Catalog No. AAB2246806
Click to view available options
Quantity:
1 g
5 g
25 g

CAS: 5751-80-4 | C7H8O2S | 156.199 g/mol

Ethyl thiophene-3-carboxylate is used as an intermediate in organic syntheses

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl thiophene-3-carboxylate is used as an intermediate in organic syntheses

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5751-80-4
Molecular Formula C7H8O2S
Molecular Weight (g/mol) 156.199
MDL Number MFCD02179226
InChI Key OYSLMAQEMAJMCL-UHFFFAOYSA-N
Synonym ethyl 3-thenoate, ethyl 3-thiophenecarboxylate, ethylthiophene-3-carboxylate, 3-ethoxycarbonyl thiophene, 3-thiophenecarboxylic acid ethyl ester, 3-thiophenecarboxylic acid, ethyl ester, thiophene-3-carboxylic acid ethyl ester, pubchem16116, ksc493o3n
PubChem CID 79824
IUPAC Name ethyl thiophene-3-carboxylate
SMILES CCOC(=O)C1=CSC=C1

Specifications

Boiling Point 207°C to 208°C
Refractive Index 1.5225
Quantity 5 g
Solubility Information Soluble in water.
Formula Weight 156.2
Percent Purity 98%
Chemical Name or Material Ethyl thiophene-3-carboxylate
EINECSNumber 227-275-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.