Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl tiglate, 98%

Catalog No. p-7081858
Click to view available options
Quantity:
25 g
100 g

CAS: 5837-78-5 | C7H12O2 | 128.171 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5837-78-5
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.171
MDL Number MFCD00015183
InChI Key OAPHLAAOJMTMLY-GQCTYLIASA-N
Synonym ethyl tiglate, tiglic acid, ethyl ester, ethyl 2-methylcrotonate, ethyl alpha-methylcrotonate, ethyl trans-2-methyl-2-butenoate, e-2-methyl-2-butenoic acid ethyl ester, ethyl e-2-methylcrotonate, ethyl trans-2-methylcrotonate, tiglic acid ethyl ester, ethyl 2-methylbut-2-enoate
PubChem CID 5281163
ChEBI CHEBI:4892
IUPAC Name ethyl (E)-2-methylbut-2-enoate
SMILES CCOC(=O)C(=CC)C

Specifications

Density 0.93
Boiling Point 156°C
Flash Point 46°C (114°F)
Refractive Index 1.434
Linear Formula CH3CH=C(CH3)CO2CH2CH3
Quantity 25 g
UN Number UN3272
Beilstein 1720895
Merck Index 14,9433
Formula Weight 128.17
Percent Purity 98%
Chemical Name or Material Ethyl tiglate
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber 227-425-6
RTECSNumber EM9252700
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.