Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Trifluoromethanesulfonate 98.0+%, TCI America™
SDP

Catalog No. T119125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T119125G 25 g
T11915G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T119125G Supplier TCI America Supplier No. T119125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 425-75-2
Molecular Formula C3H5F3O3S
Molecular Weight (g/mol) 178.125
MDL Number MFCD00000410
InChI Key UVECLJDRPFNRRQ-UHFFFAOYSA-N
Synonym ethyl triflate, ethyl trifluoromethanesulphonate, trifluoromethanesulfonic acid ethyl ester, methanesulfonic acid, trifluoro-, ethyl ester, methanesulfonic acid, 1,1,1-trifluoro-, ethyl ester, ethyl trifluoromethyl sulfonate, ethyl triflate∼trifluoromethanesulphonic acid ethyl ester, ethyl trifiate, etotf, acmc-209jqf
PubChem CID 67924
IUPAC Name ethyl trifluoromethanesulfonate
SMILES CCOS(=O)(=O)C(F)(F)F

Specifications

Color Yellow
Boiling Point 115°C
Quantity 25 g
UN Number 2924
Formula Weight 178.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Trifluoromethanesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.