Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl vinyl ether, 99%, stab.

Catalog No. p-7022779
Click to view available options
Quantity:
500 mL

CAS: 109-92-2 | C4H8O | 72.11 g/mol

Ethyl vinyl ether is used in fragrances, lubricating oil additives and spices preparation. It acts as an intermediate for sulfadiazine. It finds application in pharmaceuticals as anesthetics and analgesics. Further, it is used as a solvent in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl vinyl ether is used in fragrances, lubricating oil additives and spices preparation. It acts as an intermediate for sulfadiazine. It finds application in pharmaceuticals as anesthetics and analgesics. Further, it is used as a solvent in organic synthesis.

Solubility
Miscible with common organic solvents. Slightly miscible with water.

Notes
Air sensitive. Incompatible with strong oxidizing agents, oxygen and acids. Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-92-2
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00009248
InChI Key FJKIXWOMBXYWOQ-UHFFFAOYSA-N
Synonym ethyl vinyl ether, ethoxyethene, ethoxyethylene, ethene, ethoxy, 1-ethoxyethene, 1-ethoxyethylene, vinyl ethyl ether, vinamar, ether, ethyl vinyl, ether, vinyl ethyl
PubChem CID 8023
IUPAC Name ethenoxyethane
SMILES CCOC=C

Specifications

Melting Point -115°C
Density 0.754
Boiling Point 33°C to 34°C
Flash Point −45°C (−49°F)
Odor Ether-like
Refractive Index 1.3765
Linear Formula CH2=CHOCH2CH3
Quantity 500 mL
UN Number UN1302
Beilstein 605351
Solubility Information Miscible with common organic solvents. Slightly miscible with water.
Formula Weight 72.11
Percent Purity 99%
Chemical Name or Material Ethyl vinyl ether, stabilized
Show More Show Less
Hazard Category H225-H336
Hazard Statement GHS H Statement
H224-H319-H335-H336
Extremely flammable liquid and vapour.
Causes serious eye irritation.
May cause respiratory irritation.
May cause drowsiness or dizziness.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: VINYL ETHYL ETHER, STABILIZED
EINECSNumber 203-718-4
RTECSNumber KO0710000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.