Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Vinyl Sulfide 93.0+%, TCI America™
SDP

Catalog No. E033525ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E033525ML 25 mL
E03355ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E033525ML Supplier TCI America Supplier No. E033525ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 627-50-9
Molecular Formula C4H8S
Molecular Weight (g/mol) 88.168
MDL Number MFCD00015180
InChI Key AFGACPRTZOCNIW-UHFFFAOYSA-N
PubChem CID 12320
IUPAC Name ethenylsulfanylethane
SMILES CCSC=C

Specifications

Color Colorless
Boiling Point 91°C
Quantity 25 mL
UN Number 1993
Formula Weight 88.17
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Vinyl Sulfide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.