Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylaluminium sesquichloride, 0.4M solution in hexane, AcroSeal™, Acros Organics

Click to view available options
Quantity:
100mL
800mL
Packaging:
AcroSeal™ Glass Bottle
12075-68-2,110-54-3
C6H15Al2Cl3
247.51
MFCD00044852
LDXMUMPEDOQULK-UHFFFAOYSA-K
ethylaluminum sesquichloride,ethylaluminium sesquichloride,sesquiethylaluminum chloride,easc,aluminum, trichlorotriethyldi,ethylaluminum sesquichioride,ethylaluminum sesquichloride solution,di-,i-chlorochlorotriethyldi-aluminum,chloro diethyl alumane; dichloro ethyl alumane
25508
chloro(diethyl)alumane;dichloro(ethyl)alumane
CC[Al](CC)Cl.CC[Al](Cl)Cl
12075-68-2,110-54-3
C6H15Al2Cl3
247.51
MFCD00044852
LDXMUMPEDOQULK-UHFFFAOYSA-K
ethylaluminum sesquichloride,ethylaluminium sesquichloride,sesquiethylaluminum chloride,easc,aluminum, trichlorotriethyldi,ethylaluminum sesquichioride,ethylaluminum sesquichloride solution,di-,i-chlorochlorotriethyldi-aluminum,chloro diethyl alumane; dichloro ethyl alumane
25508
chloro(diethyl)alumane;dichloro(ethyl)alumane
CC[Al](CC)Cl.CC[Al](Cl)Cl
Lewis acid
Specific Gravity 0.701
Flash Point -22°C
Density 0.7010g/mL
Quantity 800mL
Packaging AcroSeal™ Glass Bottle
Color Colorless
Linear Formula (C2H5)2AlCl·Cl2AlC2H5
Solubility Information Solubility in water: vigorous reaction. Other solubilities: miscible with saturated aliphatic and aromatic,hydrocarbons
Formula Weight 247.51
Physical Form Liquid
Percent Purity 14.5 to 15.5% (on Al basis)
Chemical Name or Material Ethylaluminium sesquichloride
Show More Show Less

GHS H Statement
May cause damage to organs through prolonged or repeated exposure.
May cause drowsiness or dizziness.
May be fatal if swallowed and enters airways.
Toxic to aquatic life with long lasting effects.
Suspected

GHS P Statement
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.

GHS Signal Word: Danger

EINECSNumber : 235-137-7