Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylaluminium sesquichloride, 0.4M solution in hexane, AcroSeal™

Catalog No. p-3212705
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 12075-68-2 | C6H15Al2Cl3 | 247.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12075-68-2, 110-54-3
Molecular Formula C6H15Al2Cl3
Molecular Weight (g/mol) 247.51
MDL Number MFCD00044852
InChI Key LDXMUMPEDOQULK-UHFFFAOYSA-K
Synonym ethylaluminum sesquichloride, ethylaluminium sesquichloride, sesquiethylaluminum chloride, easc, aluminum, trichlorotriethyldi, ethylaluminum sesquichioride, ethylaluminum sesquichloride solution, di-, i-chlorochlorotriethyldi-aluminum, chloro diethyl alumane; dichloro ethyl alumane
PubChem CID 25508
IUPAC Name chloro(diethyl)alumane;dichloro(ethyl)alumane
SMILES CC[Al](CC)Cl.CC[Al](Cl)Cl

Specifications

Color Colorless
Density 0.7010g/mL
Flash Point -22°C
Packaging AcroSeal™ Glass Bottle
Linear Formula (C2H5)2AlCl·Cl2AlC2H5
Quantity 800 mL
Specific Gravity 0.701
Solubility Information Solubility in water: vigorous reaction. Other solubilities: miscible with saturated aliphatic and aromatic,hydrocarbons
Formula Weight 247.51
Percent Purity 14.5 to 15.5% (on Al basis)
Physical Form Liquid
Chemical Name or Material Ethylaluminium sesquichloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause damage to organs through prolonged or repeated exposure.
May cause drowsiness or dizziness.
May be fatal if swallowed and enters airways.
Toxic to aquatic life with long lasting effects.
Suspected
Precautionary Statement GHS P Statement
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
EINECSNumber 235-137-7

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.