Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylaluminum dichloride, 1M in hexanes

Catalog No. AA41712AB
Click to view available options
Quantity:
10 mL

CAS: 563-43-9 | C2H5AlCl2 | 126.94 g/mol

Ethylaluminum dichloride is used as a catalyst component in the polyolefin industry, for the synthesis of synthetic rubbers and for the dimerisation of olefins. It is also employed as a catalyst for Friedel-Crafts acylations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 563-43-9
Molecular Formula C2H5AlCl2
Molecular Weight (g/mol) 126.94
MDL Number MFCD00000457
InChI Key UAIZDWNSWGTKFZ-UHFFFAOYSA-L
Synonym ethylaluminum dichloride, aluminum, dichloroethyl, dichloroethylaluminum, ethyldichloroaluminum, dichloromonoethylaluminum, ethylaluminium dichloride, dichloro ethyl alumane, ethyl aluminum dichloride, hsdb 317, dichloro ethyl aluminum
IUPAC Name dichloro(ethyl)alumane
SMILES CC[Al](Cl)Cl

Specifications

Color Colorless to Yellow
Density 0.729 g/mL
Boiling Point 68°C to 70°C
Flash Point -22°C (-9°F)
Quantity 10 mL
UN Number UN3394
Sensitivity Air and moisture sensitive
Solubility Information Soluble in acetone. Miscible with alcohol,chloroform,ether. Reacts with water.
Formula Weight 126.95
Physical Form Liquid
Chemical Name or Material Ethylaluminum dichloride
Show More Show Less
Hazard Category H225-H250-H260-H304-H314-H335-H336-H361f-H373
Hazard Statement GHS H Statement
H225-H250-H260-H252-H304-H361-H373-H314-H318-H336
Highly flammable liquid and vapor.
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gases which may ignite spontaneously.
Self-heating in large quantities
may catch fire.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause drowsiness or dizziness.
Precautionary Statement P201-P202-P210-P222-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P281-P302+P334-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, PYROPHORIC, WATER-REACTIVE
EINECSNumber 209-248-6
RTECSNumber BD0705000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.