Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylbenzene-d10, for NMR, packaged in ampoules, 99+ atom% D

Catalog No. ac321360010
Click to view available options
Quantity:
1 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 25837-05-2 | C8H10 | 116.229 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25837-05-2
Molecular Formula C8H10
Molecular Weight (g/mol) 116.229
MDL Number MFCD00044343
InChI Key YNQLUTRBYVCPMQ-CFTAVCBPSA-N
Synonym ethylbenzene-d10, 2h10 ethylbenzene, benzene-d5, ethyl-d5, benzene-1,2,3,4,5-d5, 6-ethyl-1,1,2,2,2-d5, 1,2,3,4,5-pentadeuterio-6-1,1,2,2,2-pentadeuterioethyl benzene, decadeuteroethylbenzene, ethylbenzene-d10, 99 atom % d, 1-2 h? ethyl 2 h? benzene, ethylbenzene d10 2000 ng/microl in methanol, benzene-1,2,3,4,5-d5,6-ethyl-1,1,2,2,2-d5
PubChem CID 117648
IUPAC Name 1,2,3,4,5-pentadeuterio-6-(1,1,2,2,2-pentadeuterioethyl)benzene
SMILES CCC1=CC=CC=C1

Specifications

Physical Form Liquid
Flash Point 15°C
Refractive Index 1.492
Color Colorless
Quantity 1 mL
Density 0.9400g/mL
Deuterium (D) 99.0% min.
Boiling Point 134.5°C
Linear Formula C6D5C2D5
Formula Weight 116.25
Grade NMR
Chemical Name or Material Ethylbenzene-d10, 99+ atom% D
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.