Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ethylbenzene Solution, NMR Reference Standard, 0.1% in chloroform-d (99.8 atom % D), NMR Tube Size: 8 mm 7 in., MilliporeSigma™ Supelco™
Click to view available options
Quantity:
10 x 1.9 mL
Specifications
Specifications
Packaging | NMR Tube Size: 8 mm 7 in. |
CAS | 100-41-4 |
Concentration | 0.1% in chloroform-d (99.8 atom% D) |
Linear Formula | C8H10 |
MDL Number | MFCD00011647 |
Quantity | 10 x 1.9 mL |
Formula Weight | 106.17 |
Grade | NMR Reference Standard |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction