Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ethylbenzene Solution, NMR Reference Standard, 0.1% in chloroform-d (99.8 atom% D), TMS 0.01%, MilliporeSigma™ Supelco™
Click to view available options
Quantity:
0.235 mL
0.7 mL
Packaging:
3 mm O.D. tube
5 mm O.D. x 8 in. tube
Description
Environmental

Specifications
Specifications
Packaging | 3 mm O.D. tube |
CAS | 100-41-4 |
Linear Formula | C8H10 |
MDL Number | MFCD00011647 |
Type | NMR Reference Standard |
Quantity | 0.235 mL |
Formula Weight | 106.17 |
Grade | NMR Reference Standard |
Concentration or Composition (by Analyte or Components) | 0.1% in chloroform-d (99.8 atom % D) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction