Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Ethylboronic acid pinacol ester, 99%

Catalog No. AAH3686806
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3686806 5 g
AAH3686803 1 g
AAH3686814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH3686806 Supplier Thermo Scientific™ Supplier No. H3686806
Add to Cart

CAS: 82954-89-0 | C8H17BO2 | 156.03 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82954-89-0
Molecular Formula C8H17BO2
Molecular Weight (g/mol) 156.03
MDL Number MFCD19441153
InChI Key NPUBDPDASOEIOA-UHFFFAOYSA-N
Synonym 2-Ethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane;
IUPAC Name 2-ethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CCB1OC(C)(C)C(C)(C)O1

Specifications

Boiling Point 79-81°C / 120mbar
Quantity 5 g
Formula Weight 156.03
Percent Purity 99%
Physical Form Liquid
ShelfLife 5 Years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.