Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylcellulose, 10 mPa.s, NF, 44-51%, Spectrum™ Chemical
SDP

Catalog No. 18602558 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail

ET110, 9004-57-3

Spectrum™ Chemical Ethylcellulose, 10 mPa.s, NF is used as a coating agent, flavoring fixative, tablet binder and filler, film-former, and as a viscosity-increasing agent. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9004-57-3
Molecular Formula C23H24N6O4
Molecular Weight (g/mol) 448.48
InChI Key ARSKJXYLLONUAJ-UHFFFAOYSA-N
IUPAC Name 2-[butyl({4-[2-(2,6-dicyano-4-nitrophenyl)diazen-1-yl]-3-methylphenyl})amino]ethyl acetate
SMILES CCCCN(CCOC(C)=O)C1=CC(C)=C(C=C1)N=NC1=C(C=C(C=C1C#N)[N+]([O-])=O)C#N

Specifications

CAS Max % 100%
Quantity 45 kg
Percent Purity 44 to 51%
Grade NF
Ignition Residue 0.5%
Loss on Drying 3%
Packaging Fiber Drum
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.