Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylcellulose, 10 mPa.s, NF, 44-51%, Spectrum™ Chemical
SDP

Catalog No. 18602555 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-555 500 g Poly Bottle
18-602-554 125 g Poly Bottle
18-602-556 2.5 kg Poly Pail
18-602-557 12 kg Fiber Drum
18-602-558 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-602-555 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. ET110500GM
Add to Cart
Add to Cart

ET110, 9004-57-3

Spectrum™ Chemical Ethylcellulose, 10 mPa.s, NF is used as a coating agent, flavoring fixative, tablet binder and filler, film-former, and as a viscosity-increasing agent. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9004-57-3
Molecular Formula C23H24N6O4
Molecular Weight (g/mol) 448.48
InChI Key ARSKJXYLLONUAJ-UHFFFAOYSA-N
IUPAC Name 2-[butyl({4-[2-(2,6-dicyano-4-nitrophenyl)diazen-1-yl]-3-methylphenyl})amino]ethyl acetate
SMILES CCCCN(CCOC(C)=O)C1=CC(C)=C(C=C1)N=NC1=C(C=C(C=C1C#N)[N+]([O-])=O)C#N

Specifications

CAS Max % 100%
Quantity 500 g
Percent Purity 44 to 51%
Grade NF
Ignition Residue 0.5%
Loss on Drying 3%
Packaging Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.